missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isonicotinoylacetate, 95%, Thermo Scientific Chemicals

Product Code. 10572754
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26377-17-3
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00094021
InChI Key PCJNYGPKMQQCPX-UHFFFAOYSA-N
Synonym ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester
PubChem CID 2735202
IUPAC Name ethyl 3-oxo-3-pyridin-4-ylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=NC=C1

Specifications

Melting Point 53°C to 59°C
Color Brown-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 193.2
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl isonicotinoylacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'