missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isonicotinoylacetate, 95%, Thermo Scientific Chemicals

Catalog Number p-3736478
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
26377-17-3
C10H11NO3
193.20
MFCD00094021
PCJNYGPKMQQCPX-UHFFFAOYSA-N
ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester
2735202
ethyl 3-oxo-3-pyridin-4-ylpropanoate
CCOC(=O)CC(=O)C1=CC=NC=C1
This item is not returnable. View return policy
26377-17-3
C10H11NO3
193.20
MFCD00094021
PCJNYGPKMQQCPX-UHFFFAOYSA-N
ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester
2735202
ethyl 3-oxo-3-pyridin-4-ylpropanoate
CCOC(=O)CC(=O)C1=CC=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 53°C to 59°C
Color Brown-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 2.5 g
Formula Weight 193.2
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl isonicotinoylacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'