missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chloroformate, 99%, AcroSeal™

Product Code. 10038834 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10038834 100 mL AcroSeal™ Glass bottle 100mL
10145777 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10038834 Supplier Thermo Scientific Acros Supplier No. 430791000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 541-41-3 | C3H5ClO2 | 108.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-41-3
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00000644
InChI Key RIFGWPKJUGCATF-UHFFFAOYSA-N
Synonym ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
PubChem CID 10928
IUPAC Name ethyl carbonochloridate
SMILES CCOC(=O)Cl

Specifications

Melting Point -81.0°C
Color White
Density 1.1350g/mL
Boiling Point 94.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. Diethyl carbonate (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula ClCO2C2H5
Refractive Index 1.3950 to 1.3960
Quantity 100 mL
Merck Index 15,3839
Specific Gravity 1.135
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in benzene, chloroform and ether, soluble in most organic solvents
Viscosity 0.46 mPa.s (20°C)
Formula Weight 108.52
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Ethyl chloroformate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.