missing translation for 'onlineSavingsMsg'
Learn More

ethyle7-hydroxy-4-oxo-4H-chromene-2-carboxylate, 97%, Thermo Scientific™

Catalog Number p-2704947
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
23866-72-0
C12H10O5
234.21
MFCD00100382
RKIMFICEWCXBCE-UHFFFAOYSA-N
ethyl 7-hydroxy-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylic acid, 7-hydroxy-4-oxo-, ethyl ester, chembl82749, 2-ethoxycarbonyl-7-hydroxychromone, ethyl 7-hydroxy-4-oxo-4h-chromen-2-carboxylate, 7-hydroxy-4-oxo-4h-1-benzopyran-2-carboxylic acid ethyl ester, 7-hydroxy-4-oxo-4h-chromene-2-carboxylic acid ethyl ester, 4h-1-benzopyran-2-carboxylicacid, 7-hydroxy-4-oxo-, ethyl ester, maybridge1_002082, 7-hydroxy-4-oxo-4h-chromen-2-carboxylic acid ethyl ester
5376802
ethyl 7-hydroxy-4-oxochromene-2-carboxylate
CCOC(=O)C1=CC(=O)C2=CC=C(O)C=C2O1
This item is not returnable. View return policy
23866-72-0
C12H10O5
234.21
MFCD00100382
RKIMFICEWCXBCE-UHFFFAOYSA-N
ethyl 7-hydroxy-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylic acid, 7-hydroxy-4-oxo-, ethyl ester, chembl82749, 2-ethoxycarbonyl-7-hydroxychromone, ethyl 7-hydroxy-4-oxo-4h-chromen-2-carboxylate, 7-hydroxy-4-oxo-4h-1-benzopyran-2-carboxylic acid ethyl ester, 7-hydroxy-4-oxo-4h-chromene-2-carboxylic acid ethyl ester, 4h-1-benzopyran-2-carboxylicacid, 7-hydroxy-4-oxo-, ethyl ester, maybridge1_002082, 7-hydroxy-4-oxo-4h-chromen-2-carboxylic acid ethyl ester
5376802
ethyl 7-hydroxy-4-oxochromene-2-carboxylate
CCOC(=O)C1=CC(=O)C2=CC=C(O)C=C2O1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 234.21
Percent Purity 97%
Chemical Name or Material Ethyl 7-hydroxy-4-oxo-4H-chromene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'