missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-nitroindole-2-carboxylate, 95%

Product Code. 10423465 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10423465 10 g Glass bottle 10g
10558142 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423465 Supplier Thermo Scientific Acros Supplier No. 424660100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16732-57-3 | C11H10N2O4 | 234.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16732-57-3
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.21
MDL Number MFCD00216477
InChI Key DVFJMQCNICEPAI-UHFFFAOYSA-N
Synonym ethyl 5-nitroindole-2-carboxylate, ethyl-5-nitroindole-2-carboxylate, 5-nitroindole-2-carboxylic acid ethyl ester, ethyl-5-nitro-1h-indole-carboxylate, 1h-indole-2-carboxylic acid, 5-nitro-, ethyl ester, 2-ethoxycarbonyl-5-nitroindole, 2-ethoxycarbonyl-5-nitro-1h-indole, labotest-bb lt00441291, pubchem3016, acmc-209dw7
PubChem CID 280312
IUPAC Name ethyl 5-nitro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O

Specifications

Melting Point 220°C to 225°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: negligible
Formula Weight 234.21
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Ethyl 5-nitroindole-2-carboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.