missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-nitroindole-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog Number 10558142
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16732-57-3
C11H10N2O4
234.21
MFCD00216477
DVFJMQCNICEPAI-UHFFFAOYSA-N
ethyl 5-nitroindole-2-carboxylate, ethyl-5-nitroindole-2-carboxylate, 5-nitroindole-2-carboxylic acid ethyl ester, ethyl-5-nitro-1h-indole-carboxylate, 1h-indole-2-carboxylic acid, 5-nitro-, ethyl ester, 2-ethoxycarbonyl-5-nitroindole, 2-ethoxycarbonyl-5-nitro-1h-indole, labotest-bb lt00441291, pubchem3016, acmc-209dw7
280312
ethyl 5-nitro-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O
This item is not returnable. View return policy
16732-57-3
C11H10N2O4
234.21
MFCD00216477
DVFJMQCNICEPAI-UHFFFAOYSA-N
ethyl 5-nitroindole-2-carboxylate, ethyl-5-nitroindole-2-carboxylate, 5-nitroindole-2-carboxylic acid ethyl ester, ethyl-5-nitro-1h-indole-carboxylate, 1h-indole-2-carboxylic acid, 5-nitro-, ethyl ester, 2-ethoxycarbonyl-5-nitroindole, 2-ethoxycarbonyl-5-nitro-1h-indole, labotest-bb lt00441291, pubchem3016, acmc-209dw7
280312
ethyl 5-nitro-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 220°C to 225°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 2.5 g
Solubility Information Solubility in water: negligible
Formula Weight 234.21
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Ethyl 5-nitroindole-2-carboxylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'