missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-amino-1-methylpyrazole-4-carboxylate, 98%, ACROS Organics™

Brand:  Acros Organics 355180010

Code : 29

Additional Details : CAS Number : 31037-02-2 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10417972

  • £20.90 / 1g


Estimated Shipment
15-03-2018
Add to basket


Specifications

Chemical Identifiers

Ethyl 5-amino-1-methylpyrazole-4-carboxylate
≥97.5%
C7H11N3O2
MFCD00051652
31037-02-2
98%
169.18

Specifications

Ethyl 5-amino-1-methylpyrazole-4-carboxylate
98° to 102°C
31037-02-2
Glass bottle
≥97.5%
C7H11N3O2
MFCD00051652
Yellow
1g
Authentic
Crystals or Crystalline Powder
98%
169.18
Videos
SDS