missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methyl-5-imidazolecarboxylate, 98%, Thermo Scientific Chemicals

Product Code. 10353672
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51605-32-4
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00005199
InChI Key VLDUBDZWWNLZCU-UHFFFAOYSA-N
Synonym ethyl 4-methyl-5-imidazolecarboxylate, ethyl 4-methyl-1h-imidazole-5-carboxylate, ethyl-4-methyl-5-imidazolecarboxylate, 1h-imidazole-5-carboxylic acid, 4-methyl-, ethyl ester, 1h-imidazole-4-carboxylic acid, 5-methyl-, ethyl ester, 4-carboethoxy-5-methylimidazole, ethyl 4-methylimidazole-5-carboxylate, ethyl 5-methyl-4-imidazolecarboxylate, 5-methyl-3h-imidazole-4-carboxylic acid ethyl ester, ethyl 5-methylimidazole-4-carboxylate
PubChem CID 99200
IUPAC Name ethyl 5-methyl-1H-imidazole-4-carboxylate
SMILES CCOC(=O)C1=C(C)NC=N1

Specifications

Melting Point 204°C to 206°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Beilstein 25, I, 534
Solubility Information Solubility in water: insoluble in water
Formula Weight 154.17
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ethyl 4-methyl-5-imidazolecarboxylate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'