missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methyl-4-pentenoate, 95%

Product Code. 10088309 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10088309 1 g Glass bottle 1g
10515470 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10088309 Supplier Thermo Scientific Acros Supplier No. 427370010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4911-54-0 | C8H14O2 | 142.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4911-54-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.2
MDL Number MFCD00075160
InChI Key RRTBSPBUHUUTHR-UHFFFAOYSA-N
Synonym ethyl 4-methyl-4-pentenoate, 4-methyl-pent-4-enoic acid ethyl ester, 4-pentenoic acid,4-methyl-, ethyl ester, acmc-20akyd
PubChem CID 3416433
IUPAC Name ethyl 4-methylpent-4-enoate
SMILES CCOC(=O)CCC(=C)C

Specifications

Density 0.8910g/mL
Boiling Point 85.0°C (20.0 mmHg)
Flash Point 47°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH2=C(CH3)CH2CH2C(O)OCH2CH3
Quantity 1 g
Specific Gravity 0.891
Formula Weight 142.2
Percent Purity 95%
Chemical Name or Material Ethyl 4-methyl-4-pentenoate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl 4-methyl-4-pentenoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.