missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10633233
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10633233 10 g Amber Glass Bottle 10g
10283712 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10633233 Supplier Thermo Scientific Maybridge Supplier No. SPB03021EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-03-9
Molecular Formula C14H12F3NO2S
Molecular Weight (g/mol) 315.31
MDL Number MFCD00068104
InChI Key LPIXRQSYBTUXOQ-UHFFFAOYSA-N
Synonym ethyl 4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carboxylate, 4-methyl-2-4-trifluoromethylphenyl thiazole-5-carboxylic acid ethyl ester, ethyl 4-methyl-2-4-trifluoromethyl phenyl-thiazole-5-carboxylate, 4-methyl-2-4-trifluoromethyl-phenyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 4-methyl-2-4-trifluoromethyl phenyl-, ethyl ester, ethyl4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carboxylate, ethyl 4-methyl-2-4-trifluoromethyl phenyl thiazo
PubChem CID 2737210
IUPAC Name ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)C2=CC=C(C=C2)C(F)(F)F)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 315.31
Percent Purity 97%
Chemical Name or Material Ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.