missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-fluorobenzoate, 99%

Product Code. 10086293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10086293 100 g Glass bottle 100g
10615582 5 g Glass bottle 5g
10523311 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10086293 Supplier Thermo Scientific Acros Supplier No. 146801000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 451-46-7 | C9H9FO2 | 168.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 451-46-7
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD00000351
InChI Key UMPRJGKLMUDRHL-UHFFFAOYSA-N
Synonym 4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate
PubChem CID 67976
IUPAC Name ethyl 4-fluorobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)F

Specifications

Melting Point 25°C to 27°C
Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 210°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CO2C2H5
Refractive Index 1.4850 to 1.487
Quantity 100 g
Beilstein 09, 334
Merck Index 10, 4076
Specific Gravity 1.14
Solubility Information Solubility in water: insoluble
Formula Weight 168.17
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Ethyl 4-fluorobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-fluorobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.