missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-fluorobenzoate, 99%, Thermo Scientific Chemicals

Product Code. 10523311
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 451-46-7
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD00000351
InChI Key UMPRJGKLMUDRHL-UHFFFAOYSA-N
Synonym 4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate
PubChem CID 67976
IUPAC Name ethyl 4-fluorobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)F

Specifications

Melting Point 25°C to 27°C
Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 210°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CO2C2H5
Refractive Index 1.4850 to 1.487
Quantity 25 g
Beilstein 09, 334
Merck Index 10, 4076
Specific Gravity 1.14
Solubility Information Solubility in water: insoluble
Formula Weight 168.17
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Ethyl 4-fluorobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-fluorobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'