missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10031934
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 10031934

Brand: Thermo Scientific Maybridge SPB04750EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 720-01-4
Molecular Formula C8H6ClF3N2O2
Molecular Weight (g/mol) 254.59
MDL Number MFCD00173897
InChI Key DSULCDCBENGHAX-UHFFFAOYSA-N
Synonym ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 4-chloro-2-trifluoromethyl-, ethyl ester, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylicacidethylester, ethyl 2-trifluoromethyl-4-chloropyrimidine-5-carboxylate, ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-car, ethyl 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylate, ethyl-4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-chloro-2-trifluoromethyl-, ethyl ester
PubChem CID 2737166
IUPAC Name ethyl 4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1Cl)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 254.59
Percent Purity 97%
Chemical Name or Material ethyle4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.