missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10180942
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10180942 1 g 1g
10096724 5 g 5g
10031934 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10180942 Supplier Thermo Scientific Maybridge Supplier No. SPB04750DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 720-01-4
Molecular Formula C8H6ClF3N2O2
Molecular Weight (g/mol) 254.59
MDL Number MFCD00173897
InChI Key DSULCDCBENGHAX-UHFFFAOYSA-N
Synonym ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 4-chloro-2-trifluoromethyl-, ethyl ester, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylicacidethylester, ethyl 2-trifluoromethyl-4-chloropyrimidine-5-carboxylate, ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-car, ethyl 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylate, ethyl-4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-chloro-2-trifluoromethyl-, ethyl ester
PubChem CID 2737166
IUPAC Name ethyl 4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1Cl)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 254.59
Percent Purity 97%
Chemical Name or Material ethyle4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.