missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetylbenzoate, 99%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
This item is not returnable. View return policy
38430-55-6
C11H12O3
192.21
MFCD00013241
GLOAPLPTWAXAIG-UHFFFAOYSA-N
4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
600911
ethyl 4-acetylbenzoate
CCOC(=O)C1=CC=C(C=C1)C(C)=O
This item is not returnable. View return policy
38430-55-6
C11H12O3
192.21
MFCD00013241
GLOAPLPTWAXAIG-UHFFFAOYSA-N
4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
600911
ethyl 4-acetylbenzoate
CCOC(=O)C1=CC=C(C=C1)C(C)=O
Color White to Beige
Melting Point 50°C to 55°C
Assay Percent Range 98.5% min. (GC)
Infrared Spectrum Authentic
Linear Formula CH3CH2OOCC6H4COCH3
Packaging Glass bottle
Quantity 1g
Formula Weight 192.21
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Ethyl 4-acetylbenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-acetylbenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only