missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetylbenzoate, 99%, ACROS Organics™

£52.14 - £178.00

Chemical Identifiers

CAS 38430-55-6
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00013241
InChI Key GLOAPLPTWAXAIG-UHFFFAOYSA-N
Synonym 4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
PubChem CID 600911
IUPAC Name ethyl 4-acetylbenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C(=O)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10001554
SDS View Documents
Acros Organics
397310010
1g Glass bottle
£52.14
1g
View alternative products
Save Up to 
»
Estimated Shipment
29-01-2021
Please to purchase this item. Need a web account? Register with us today!
10725125
SDS View Documents
Acros Organics
397310050
5g Glass Bottle
£178.00
5g
View alternative products
Save Up to 
»
Estimated Shipment
03-02-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

38430-55-6
192.21
GLOAPLPTWAXAIG-UHFFFAOYSA-N
600911
CCOC(=O)C1=CC=C(C=C1)C(=O)C
C11H12O3
MFCD00013241
4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
ethyl 4-acetylbenzoate

Specifications

Ethyl 4-acetylbenzoate
Glass bottle
50°C to 55°C
38430-55-6
C11H12O3
MFCD00013241
GLOAPLPTWAXAIG-UHFFFAOYSA-N
ethyl 4-acetylbenzoate
600911
Crystalline Powder
Authentic
White to Beige
1g
98.5% min. (GC)
CH3CH2OOCC6H4COCH3
4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
CCOC(=O)C1=CC=C(C=C1)C(=O)C
192.21
192.21
99%
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl 4-acetylbenzoate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS