missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-[(2-ethoxy-2-oxoethyl)thio]butanoate, 97%, Thermo Scientific™

Product Code. 10374712
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10374712 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10374712 Supplier Thermo Scientific Maybridge Supplier No. KM04026DA
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 63449-37-6
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H18O4S
Linear Formula CH3CH2O2C(CH2)3SCH2CO2CH2CH3
MDL Number MFCD00085074
Quantity 1 g
Synonym ethyl 4-2-ethoxy-2-oxoethyl thio butanoate, ethyl 4-2-ethoxy-2-oxoethyl sulfanyl butanoate, ethyl 4-ethoxycarbonyl methylthio butanoate, ethyl 4-2-ethoxy-2-oxoethyl thio butyrate, ethyl 4-2-ethoxy-2-oxoethyl sulfanylbutanoate, ethyl 4-2-ethoxy-2-oxoethyl sulphanyl butanoate, butanoic acid,4-2-ethoxy-2-oxoethyl thio-,ethyl ester, butanoic acid, 4-2-ethoxy-2-oxoethyl thio-, ethyl ester
InChI Key QAUACNHILIEKFW-UHFFFAOYSA-N
SMILES CCOC(=O)CCCSCC(=O)OCC
IUPAC Name ethyl 4-(2-ethoxy-2-oxoethyl)sulfanylbutanoate
Molecular Weight (g/mol) 234.31
PubChem CID 2778847
Formula Weight 234.31
Percent Purity 97%
Chemical Name or Material ethyle4-[(2-ethoxy-2-oxoethyl)thio]butanoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.