missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-isocyanatopropionate, 98%, Acros Organics™

Brand:  Acros Organics 319270010

Code : 29

Additional Details : CAS Number : 5100-34-5 Weight : 0.05100kg Transport : UN number : 2206 Chem class : 2 Pack group : III

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10319320

  • £28.00 / 1g


Estimated Shipment
12-02-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 3-isocyanatopropionate
98%
OCNCH2CH2C(=O)OCH2CH3
MFCD00078461
98%
C6H9NO3
143.07

Specifications

Ethyl 3-isocyanatopropionate
Clear colorless to yellow
5100-34-5
Authentic
1.4275 to 1.4295
Liquid
98%
OCNCH2CH2C(=O)OCH2CH3
MFCD00078461
68.0°C (2.0 mmHg)
1g
1.0870g/mL
Glass bottle
1.087
98%
C6H9NO3
143.07
Videos
SDS