missing translation for 'onlineSavingsMsg'
Learn More

ethyle3,5-dichlorobenzoate, 97%, Thermo Scientific™

Product Code. 10039521
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10039521 10 g Amber Glass Bottle 10g
10029521 1 g Amber Glass Bottle 1g
10138784 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10039521 Supplier Thermo Scientific Maybridge Supplier No. TL00759EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 91085-56-2
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.06
MDL Number MFCD00221467
InChI Key JRLNLVFPSMDPLU-UHFFFAOYSA-N
Synonym ethyl3,5-dichlorobenzoate, 3,5-dichlorobenzoic acid ethyl ester, rarechem al bi 0057, 5-ethoxycarbonyl-1,3-dichlorobenzene, benzoic acid, 3,5-dichloro-, ethyl ester, pubchem10323, ethyl 3,5-dichloro benzoate, ethyl 3,5-bis chloranyl benzoate, 3,5-dichlorobenzoic acid ethyl etster
PubChem CID 2736404
IUPAC Name ethyl 3,5-dichlorobenzoate
SMILES CCOC(=O)C1=CC(Cl)=CC(Cl)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3C(O)OCH2CH3
Quantity 10 g
Formula Weight 219.07
Percent Purity 97%
Chemical Name or Material Ethyl 3, 5-dichlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.