missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle3,5-dichlorobenzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | Cl2C6H3C(O)OCH2CH3 |
Quantity | 25 g |
Formula Weight | 219.07 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 3, 5-dichlorobenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'