Learn More
Thermo Scientific Maybridge Ethyl 3-(4-hydroxyphenyl)propanoate, 1g, 97%
Brand: Thermo Scientific Maybridge BTB00914DA
Ethyl 3-(4-hydroxyphenyl)propanoate, 1g, 97%
Code : 31
Additional Details : CAS Number : 23795-02-0 Weight : 0.05100kg