missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-mercapto-1H-imidazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10272982
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10272982 10 g Amber Glass Bottle 10g
10553242 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10272982 Supplier Thermo Scientific Maybridge Supplier No. KM06163EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 64038-64-8
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00068042
InChI Key PONOGPICUOALRU-UHFFFAOYSA-N
Synonym ethyl 2-mercapto-1h-imidazole-4-carboxylate, 4-ethoxycarbonylimidazole-2-thiol, usaf el-63, imidazole-2-thiol, 5-ethoxycarbonyl, 2-mercapto-5-carbethoxyimidazole, imidazole, 2-mercapto-5-carbethoxy, ethyl 2-sulfanyl-1h-imidazole-4-carboxylate, 2-mercapto-1h-imidazole-4-carboxylic acid ethyl ester, ethyl 2-mercaptoimidazole-4-carboxylate, maybridge1_005066
PubChem CID 2736338
IUPAC Name ethyl 2-sulfanylidene-1,3-dihydroimidazole-4-carboxylate
SMILES CCOC(=O)C1=CNC(=S)N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 172.21
Percent Purity 97%
Chemical Name or Material Ethyl 2-mercapto-1H-imidazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.