missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-chloro-3-cyano-6-methylisonicotinate, 97%, Thermo Scientific™

Product Code. 10563242
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10563242 1 g Amber Glass Bottle 1g
10405712 250 mg Amber Glass Bottle 250mg
10192834 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10563242 Supplier Thermo Scientific Maybridge Supplier No. RF00966DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 40108-12-1
Molecular Formula C10H9ClN2O2
Molecular Weight (g/mol) 224.644
MDL Number MFCD00023416
InChI Key XUXBHWPRQGVLRY-UHFFFAOYSA-N
Synonym ethyl 2-chloro-3-cyano-6-methylisonicotinate, ethyl-2-chloro-3-cyano-6-methylisonicotinate, isonicotinic acid, 2-chloro-3-cyano-6-methyl-, ethyl ester, 2-chloro-3-cyano-6-methylisonicotinic acid ethyl ester, 2-chloro-3-cyano-4-carbethoxy-6-methylpyridine, ethyl2-chloro-3-cyano-6-methylisonicotinate, 2-chloro-3-cyano-6-methyl-isonicotinic acid ethyl ester, 4-pyridinecarboxylic acid, 2-chloro-3-cyano-6-methyl-, ethyl ester, 4-pyridinecarboxylicacid, 2-chloro-3-cyano-6-methyl-, ethyl ester
PubChem CID 142408
IUPAC Name ethyl 2-chloro-3-cyano-6-methylpyridine-4-carboxylate
SMILES CCOC(=O)C1=CC(=NC(=C1C#N)Cl)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 224.65
Percent Purity 97%
Chemical Name or Material ethyle2-chloro-3-cyano-6-methylisonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.