missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromobenzoate, 98%, Thermo Scientific Chemicals

Product Code. 10321574 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10321574 100 g Glass bottle 100g
10761341 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10321574 Supplier Thermo Scientific Acros Supplier No. 308531000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6091-64-1
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00015443
InChI Key BIHHBTVQFPVSTE-UHFFFAOYSA-N
Synonym 2-bromobenzoic acid ethyl ester, benzoic acid, 2-bromo-, ethyl ester, ethyl-2-bromobenzoate, ethyl2-bromobenzoate, ethyl bromobenzoate, ethyl o-bromobenzoate, pubchem3937, brombenzoesaureathylester, acmc-209mn8, ksc493o4r
PubChem CID 80186
IUPAC Name ethyl 2-bromobenzoate
SMILES CCOC(=O)C1=CC=CC=C1Br

Specifications

Color Yellow
Density 1.4400g/mL
Boiling Point 117°C (17.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC6H4CO2C2H5
Refractive Index 1.5430 to 1.545
Quantity 100 g
Specific Gravity 1.44
Formula Weight 229.07
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 2-bromobenzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.