missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-amino-5-methyl-4-phenylthiophene-3-carboxylate, 97%, Thermo Scientific™

Product Code. p-2704095
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10180912

Brand: Thermo Scientific Maybridge AW00937DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4815-37-6
Molecular Formula C14H15NO2S
Molecular Weight (g/mol) 261.339
MDL Number MFCD01763475
InChI Key SXONDAWSQJWZEO-UHFFFAOYSA-N
Synonym 2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, ethyl ester, maybridge1_000503, upcmld-dp080, upcmld-dp080:001, ethyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 3-thiophenecarboxylic acid,2-amino-5-methyl-4-phen, 2-amino-4-phenyl-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-4-phenyl-, ethyl ester
PubChem CID 693902
IUPAC Name ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC(=C1C2=CC=CC=C2)C)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 261.34
Percent Purity 97%
Chemical Name or Material ethyle2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.