missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(4-chlorophenoxy)-2-methylpropanoate, 97%, Maybridge

Product Code. 10158504
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10158504 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10158504 Supplier Thermo Scientific Maybridge Supplier No. RDP00167EB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 637-07-0
CAS Min % 97.0
Density 1.1370g/mL
Assay Percent Range 97%
Molecular Formula C12H15ClO3
Linear Formula ClC6H4OC(CH3)2C(O)OCH2CH3
MDL Number MFCD00000615
Quantity 25 g
Synonym clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
InChI Key KNHUKKLJHYUCFP-UHFFFAOYSA-N
SMILES CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
IUPAC Name ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
Molecular Weight (g/mol) 242.70
PubChem CID 2796
ChEBI CHEBI:3750
Formula Weight 242.7
Percent Purity 97%
Chemical Name or Material Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.