missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate, 97%, Thermo Scientific™

Product Code. 10190602
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10190602 50 g 50g
10223092 1 g 1g
10485712 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10190602 Supplier Thermo Scientific Maybridge Supplier No. S12083EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 61713-23-3
Molecular Formula C9H16O4S2
Molecular Weight (g/mol) 252.343
MDL Number MFCD00026901
InChI Key NBNGVIRHOBZBHM-UHFFFAOYSA-N
Synonym methylenebis ethyl thioglycolate, ethyl 2-2-ethoxy-2-oxoethyl thio methyl thio acetate, ethyl 2-2-ethoxy-2-oxoethyl sulfanyl methyl sulfanyl acetate, ethyl 2-ethoxy-2-oxoethyl sulfanyl methyl sulfanyl acetate, diethyl 2,2'-methylenedisulfanediyl diacetate, diethyl 2,2'-methylenebis sulfanediyl diacetate, ethyl 2-ethoxycarbonyl methylthio methylthio acetate, ethyl 2-2-ethoxy-2-oxoethyl sulfanylmethylsulfanyl acetate, aceticacid, 2,2'-methylenebis thio bis-, diethyl ester 9ci
PubChem CID 521987
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethyl)sulfanylmethylsulfanyl]acetate
SMILES CCOC(=O)CSCSCC(=O)OCC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CH3CH2O2CCH2SCH2SCH2CO2CH2CH3
Quantity 50 g
Formula Weight 252.35
Percent Purity 97%
Chemical Name or Material Ethyl 2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.