missing translation for 'onlineSavingsMsg'
Learn More

ethyle1-(4-cyanophenyl)-4-piperidinecarboxylate, 97%, Thermo Scientific™

Product Code. p-2702940
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10742303 1 g Amber Glass Bottle 1g
10314532 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10742303 Supplier Thermo Scientific Maybridge Supplier No. MO00017DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 352018-90-7
Molecular Formula C15H18N2O2
Molecular Weight (g/mol) 258.32
MDL Number MFCD03086198
InChI Key LIWHXOHASZYDEM-UHFFFAOYSA-N
Synonym ethyl 1-4-cyanophenyl piperidine-4-carboxylate, ethyl 1-4-cyanophenyl-4-piperidinecarboxylate, 4-4-ethoxycarbonyl-piperidino-benzonitrile, ethyl 1-cyanophenyl-4-piperidinecarboxylate, ethyl 1-4-cyanophenyl-piperidine-4-carboxylate, 1-4-cyanophenyl piperidine-4-carboxylic acid ethyl ester, 4-piperidinecarboxylicacid, 1-4-cyanophenyl-, ethyl ester
PubChem CID 2779638
IUPAC Name ethyl 1-(4-cyanophenyl)piperidine-4-carboxylate
SMILES CCOC(=O)C1CCN(CC1)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 258.32
Percent Purity 97%
Chemical Name or Material Ethyl 1-(4-cyanophenyl)-4-piperidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.