missing translation for 'onlineSavingsMsg'
Learn More

ethyle1-(3-aminobenzyl)piperidine-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10121282
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10121282 1 g 1g
10753137 5 g 5g
10482644 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10121282 Supplier Thermo Scientific Maybridge Supplier No. SPB08422DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306937-22-4
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.353
MDL Number MFCD02180894
InChI Key WXYBQSXOZUFNJY-UHFFFAOYSA-N
PubChem CID 2800723
IUPAC Name ethyl 1-[(3-aminophenyl)methyl]piperidine-4-carboxylate
SMILES CCOC(=O)C1CCN(CC1)CC2=CC(=CC=C2)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym ethyl 1-3-aminobenzyl piperidine-4-carboxylate, ethyl 1-3-aminophenyl methyl piperidine-4-carboxylate, 4-piperidinecarboxylicacid, 1-3-aminophenyl methyl-, ethyl ester, ethyl1-3-aminobenzyl piperidine-4-carboxylate, 1-3-aminobenzyl-4-ethoxycarbonyl piperidine, 3-4-ethoxycarbonyl piperidin-1-yl methyl aniline, 1-3-aminophenyl methyl-4-piperidinecarboxylic acid ethyl ester
Formula Weight 262.35
Percent Purity 97%
Chemical Name or Material ethyle1-(3-aminobenzyl)piperidine-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.