missing translation for 'onlineSavingsMsg'
Learn More

Ethylcyclohexane, 99+%, Thermo Scientific Chemicals

Product Code. p-128998 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10550751

Brand: Thermo Scientific Acros 118310250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1678-91-7
Molecular Formula C8H16
Molecular Weight (g/mol) 112.21
MDL Number MFCD00001523
InChI Key IIEWJVIFRVWJOD-UHFFFAOYSA-N
Synonym cyclohexane, ethyl, ethyl cyclohexane, cyclohexylethane, ethyl-cyclohexane, acmc-1c1qu, ethylcyclohexane, dsstox_cid_30334, dsstox_gsid_51779, wln: l6tj a2, qspl 100
PubChem CID 15504
IUPAC Name ethylcyclohexane
SMILES CCC1CCCCC1

Specifications

Melting Point -111°C
Color Colorless
Density 0.7800g/mL
Boiling Point 130°C to 132°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H11C2H5
Refractive Index 1.4311 to 1.4331
Quantity 25 mL
Beilstein 05, 35
Specific Gravity 0.78
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,carbon tetrachloride,lignoin
Formula Weight 112.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethylcyclohexane, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylcyclohexane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.