missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, puriss. p.a., ≥99.0% (GC), Honeywell™ Riedel-de-Haën™

Product Code. 15640660
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 15640660

Brand: Honeywell Riedel-de Haën 030801L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 100-41-4
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00011647
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1

Specifications

Density 0.867g/mL at 25°C
Flash Point 15°C (59°F)
Linear Formula C6H5C2H5
Quantity 1 L
UN Number UN1175
Beilstein 1901871
Formula Weight 106.17g/mol
Percent Purity ≥99.0% (GC)
Grade puriss. p.a.
Chemical Name or Material Ethylbenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.