missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D

Product Code. 10175900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10175900 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10175900 Supplier Thermo Scientific Acros Supplier No. 321360010
  • ONLINE EXCLUSIVE PRICE
    £43.30
    £39.80 / 1mL
    Save £3.50
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 25837-05-2 | C8H10 | 116.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 0.9400g/mL
Deuterium (D) 99.0% min.
Flash Point 15°C
Refractive Index 1.492
Quantity 1 mL
CAS 25837-05-2
Boiling Point 134.5°C
Molecular Formula C8H10
Linear Formula C6D5C2D5
MDL Number MFCD00044343
Synonym ethylbenzene-d10, 2h10 ethylbenzene, benzene-d5, ethyl-d5, benzene-1,2,3,4,5-d5, 6-ethyl-1,1,2,2,2-d5, 1,2,3,4,5-pentadeuterio-6-1,1,2,2,2-pentadeuterioethyl benzene, decadeuteroethylbenzene, ethylbenzene-d10, 99 atom % d, 1-2 h? ethyl 2 h? benzene, ethylbenzene d10 2000 ng/microl in methanol, benzene-1,2,3,4,5-d5,6-ethyl-1,1,2,2,2-d5
InChI Key YNQLUTRBYVCPMQ-CFTAVCBPSA-N
SMILES CCC1=CC=CC=C1
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,1,2,2,2-pentadeuterioethyl)benzene
Molecular Weight (g/mol) 116.229
PubChem CID 117648
Formula Weight 116.25
Grade NMR
Chemical Name or Material Ethylbenzene-d10, 99+ atom% D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylbenzene-d10
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.