missing translation for 'onlineSavingsMsg'
Learn More

Ethylamine Solution (2.0 M in THF), TRC

Product Code. 30370232
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370232 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370232 Supplier Toronto Research Chemicals Supplier No. TRCE9456505ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylamine
CAS 75-04-7
Molecular Formula C2 H7 N
Quantity 5 mL
Synonym Ethylamine, 1-Aminoethane, 45: PN: WO2023274347 PAGE: 25 claimed sequence, Aminoethane, Monoethylamine, n-Ethylamine, R 631
InChI Formula InChI=1S/C2H7N/c1-2-3/h2-3H2,1H3
SMILES CCN
IUPAC Name ethanamine
Molecular Weight (g/mol) 45.08
Formula Weight 45.0578
Concentration or Composition (by Analyte or Components) 2 M
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.