missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylamine Hydrochloride, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Ethylamine Hydrochloride |
| CAS | 557-66-4 |
| Molecular Formula | C2H8ClN |
| Quantity | 100 g |
| Synonym | Ethylammonium Chloride, Monoethylammonium Chloride, N-Ethylammonium Chloride |
| InChI Formula | InChI=1S/C2H7N.ClH/c1-2-3;/h2-3H2,1H3;1H |
| SMILES | NCC.Cl |
| Molecular Weight (g/mol) | 81.54 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction