missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Violet (Technical Grade), TRC

Product Code. 30384171
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30384171

Brand: Toronto Research Chemicals TRCE937530250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Violet
CAS 2390-59-2
Molecular Formula C31 H42 N3 . Cl
Quantity 250 mg
Synonym C.I. Basic Violet 4 (7CI,8CI), Ethanaminium, N-[4-[bis[4-(diethylamino)phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-, chloride (1:1), Ethanaminium, N-[4-[bis[4-(diethylamino)phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-, chloride (9CI), Ethyl violet (6CI), Basic Violet 4, C.I. 42600, Ethyl Violet AX, Ethyl Violet GGA, Ethyl crystal violet, Lowacryl Violet 4, Shikiso Acid Brilliant Blue 6B, Tris(p-(diethylamino)phenyl)methylium chloride
InChI Formula InChI=1S/C31H42N3.ClH/c1-7-32(8-2)28-19-13-25(14-20-28)31(26-15-21-29(22-16-26)33(9-3)10-4)27-17-23-30(24-18-27)34(11-5)12-6;/h13-24H,7-12H2,1-6H3;1H/q+1;/p-1
SMILES [Cl-].CCN(CC)c1ccc(cc1)C(=C2C=CC(=[N+](CC)CC)C=C2)c3ccc(cc3)N(CC)CC
IUPAC Name [4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride
Molecular Weight (g/mol) 492.1383
Formula Weight 491.3067
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.