missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Violet, 80%, Thermo Scientific™

Product Code. 10279740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10279740 25 g 25g
15535096 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10279740 Supplier Thermo Scientific Acros Supplier No. 410490250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2390-59-2
Molecular Formula C31H42ClN3
Molecular Weight (g/mol) 492.15
MDL Number MFCD00011765
InChI Key JVICFMRAVNKDOE-UHFFFAOYSA-M
Synonym Basic Violet 4, C.I. 42600
PubChem CID 16955
IUPAC Name [4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC

Specifications

Dye Content 75% min.
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol and ethanol
Infrared Spectrum Authentic
Lambda Max 593 to 599nm (c=0.1, H2O)
Formula Weight 492.14
Loss on Drying 4% max.
Percent Purity 80%
Packaging Glass bottle
Quantity 25 g
Chemical Name or Material Ethyl Violet

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.