missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Vinyllactate-13D2,d3, TRC

Product Code. 30382698
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.5mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382698 0.5 mg 0.5mg
30378881 2.5 mg 2.5mg
30384208 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30382698 Supplier Toronto Research Chemicals Supplier No. TRCE9321070.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Vinyllactate-13C2,d3
Molecular Formula 13C2 C5 D3 H9 O3
Quantity 0.5 mg
Synonym Ethyl 2-Vinyllactate-13C2,d3
InChI Formula InChI=1S/C7H12O3/c1-4-7(3,9)6(8)10-5-2/h4,9H,1,5H2,2-3H3/i3D3,6+1,7+1
SMILES [2H]C([2H])([2H])[13C](O)(C=C)[13C](=O)OCC
IUPAC Name ethyl 2-hydroxy-2-(trideuteriomethyl)(1,2-13C2)but-3-enoate
Molecular Weight (g/mol) 149.172
Formula Weight 149.104
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.