missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Vinyl Ether, TRC

Product Code. 30383234
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30383234 100 mL -
30374789 2.5 L 2.5 L
30372853 1 L -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383234 Supplier Toronto Research Chemicals Supplier No. TRCE931960100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-92-2
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
Synonym 1-Ethoxyethene, 1-Ethoxyethylene, EVE, Ethoxyethene, Ethoxyethylene, Ethyl Vinyl Ether, Ethyloxyethene, NSC 8405, Vinamar, Vinyl Ethyl Ether, [(Ethenyl)oxy]ethane, [(Vinyl)oxy]ethane
SMILES CCOC=C

Specifications

Quantity 100 mL
InChI Formula InChI=1S/C4H8O/c1-3-5-4-2/h3H,1,4H2,2H3
Formula Weight 72.0575
Chemical Name or Material Ethyl Vinyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.