missing translation for 'onlineSavingsMsg'
Learn More

Ethyl valerate, 99%

Product Code. 10156940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10156940 5 g Glass bottle 5g
10065660 100 g Glass bottle 100g
10474125 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10156940 Supplier Thermo Scientific Acros Supplier No. 410470050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 539-82-2 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-82-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009479
InChI Key ICMAFTSLXCXHRK-UHFFFAOYSA-N
Synonym ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462
PubChem CID 10882
IUPAC Name ethyl pentanoate
SMILES CCCCC(=O)OCC

Specifications

Color Colorless
Density 0.8700g/mL
Boiling Point 145°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)3COOC2H5
Refractive Index 1.3990 to 1.401
Quantity 5 g
Merck Index 15, 10090
Specific Gravity 0.87
Formula Weight 130.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl valerate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl valerate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.