missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trimethylacetopyruvate, 98%

Product Code. 11472158
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472158 5 g 5g
11462158 1 g 1g
11482158 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472158 Supplier Thermo Scientific Alfa Aesar Supplier No. A15164.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13395-36-3 | C10H16O4 | 200.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13395-36-3
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00052319
InChI Key NIMKIMUBJFWPTD-UHFFFAOYSA-N
Synonym ethyl trimethylacetopyruvate, hexanoic acid, 5,5-dimethyl-2,4-dioxo-, ethyl ester, 5,5-dimethyl-2,4-dioxo-hexanoic acid ethyl ester, 5,5-dimethyl-2,4-dioxohexanoic acid ethyl ester, ethyl pivaloylpyruvate, pivaloylpyruvic acid ethyl ester, 3-pivaloylpyruvic acid ethyl ester, ethyl5,5-dimethyl-2,4-dioxohexanoate, trimethylacetopyruvic acid ethyl ester, ethyl 5,5-dimethyl-2,4-dioxo-hexanoate
PubChem CID 83403
IUPAC Name ethyl 5,5-dimethyl-2,4-dioxohexanoate
SMILES CCOC(=O)C(=O)CC(=O)C(C)(C)C

Specifications

Boiling Point 68°C to 70°C (0.5 mmHg)
Refractive Index 1.47
Quantity 5 g
Beilstein 510374
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material Ethyl trimethylacetopyruvate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.