missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trimethylacetate, 99%

Product Code. 11454434
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454434 100 g 100g
11444434 25 g 25g
11464434 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454434 Supplier Thermo Scientific Alfa Aesar Supplier No. A13100.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3938-95-2 | C7H14O2 | 130.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3938-95-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009129
InChI Key HHEIMYAXCOIQCJ-UHFFFAOYSA-N
Synonym ethyl trimethylacetate, ethyl pivalate, propanoic acid, 2,2-dimethyl-, ethyl ester, ethyl 2,2-dimethylpropionate, pivalic acid, ethyl ester, ethylpivalate, trimethylacetic acid, ethyl ester, unii-738s62lzwy, ethyltrimethylacetate, 2,2-dimethylpropanoic acid ethyl ester
PubChem CID 19838
IUPAC Name ethyl 2,2-dimethylpropanoate
SMILES CCOC(=O)C(C)(C)C

Specifications

Melting Point -90°C
Density 0.891
Boiling Point 116°C to 118°C
Flash Point 16°C (60°F)
Odor Stench
Refractive Index 1.393
Quantity 100 g
UN Number UN3272
Beilstein 1747617
Merck Index 14,7511
Formula Weight 130.19
Percent Purity 99%
Chemical Name or Material Ethyl trimethylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.