missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Trifluoromethanesulfonate, TRC

Product Code. 30372525
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372525 500 mg
30368838 50 mg
30371517 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372525 Supplier Toronto Research Chemicals Supplier No. TRCE937738500MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 425-75-2
Molecular Formula C3H5F3O3S
Molecular Weight (g/mol) 178.13
SMILES CCOS(=O)(=O)C(F)(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C3H5F3O3S/c1-2-9-10(7,8)3(4,5)6/h2H2,1H3
Formula Weight 177.9911
Chemical Name or Material Ethyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.