missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoromethanesulfonate, 99%, Thermo Scientific™

Product Code. 11392126
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392126 1 g 1g
11302136 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392126 Supplier Thermo Scientific Alfa Aesar Supplier No. L03383.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl trifluoromethanesulfonate is a powerful ethylating agent.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 425-75-2
Molecular Formula C3H5F3O3S
Molecular Weight (g/mol) 178.125
MDL Number MFCD00000410
InChI Key UVECLJDRPFNRRQ-UHFFFAOYSA-N
Synonym ethyl triflate, ethyl trifluoromethanesulphonate, trifluoromethanesulfonic acid ethyl ester, methanesulfonic acid, trifluoro-, ethyl ester, methanesulfonic acid, 1,1,1-trifluoro-, ethyl ester, ethyl trifluoromethyl sulfonate, ethyl triflate∼trifluoromethanesulphonic acid ethyl ester, ethyl trifiate, etotf, acmc-209jqf
PubChem CID 67924
IUPAC Name ethyl trifluoromethanesulfonate
SMILES CCOS(=O)(=O)C(F)(F)F

Specifications

Boiling Point 115°C to 118°C
Flash Point 35°C (95°F)
Odor Pungent
Linear Formula CF3SO3CH2CH3
Quantity 1 g
UN Number UN2920
Beilstein 1770746
Formula Weight 178.13
Percent Purity 99%
Chemical Name or Material Ethyl trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.