missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoroacetoacetate, 97%

Product Code. 11454724
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454724 250 g 250g
11434724 10 g 10g
11444724 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454724 Supplier Thermo Scientific Alfa Aesar Supplier No. A13717.30
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 372-31-6 | C6H7F3O3 | 184.114 g/mol

Ethyl trifluoroacetoacetate is used as an intermediate in synthetic chemistry. It is also used as an intermediate for agro chemicals and pharmaceuticals. Further, it is used in the preparation of fluorinated 2-thiouracil analogs as antithyroid agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoroacetoacetate is used as an intermediate in synthetic chemistry. It is also used as an intermediate for agro chemicals and pharmaceuticals. Further, it is used in the preparation of fluorinated 2-thiouracil analogs as antithyroid agents.

Solubility
Miscible with water, ethanol, benzene and organic solvents.

Notes
Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-31-6
Molecular Formula C6H7F3O3
Molecular Weight (g/mol) 184.114
MDL Number MFCD00000424
InChI Key OCJKUQIPRNZDTK-UHFFFAOYSA-N
Synonym ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester
PubChem CID 67793
IUPAC Name ethyl 4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC(=O)CC(=O)C(F)(F)F

Specifications

Melting Point -30°C
Density 1.259
Boiling Point 129°C to 130°C
Flash Point 38°C (100°F)
Odor Odorless
Refractive Index 1.376
Quantity 250 g
UN Number UN3272
Beilstein 608353
Solubility Information Miscible with water,ethanol,benzene and organic solvents.
Formula Weight 184.12
Percent Purity ≥98%
Chemical Name or Material Ethyl trifluoroacetoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.