missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoroacetate, 99%

Product Code. 11424523
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424523 250 g 250g
11414523 50 g 50g
11404523 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424523 Supplier Thermo Scientific Alfa Aesar Supplier No. A11520.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 383-63-1 | C4H5F3O2 | 142.077 g/mol

Ethyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.

Solubility
Slightly miscible with water. Miscible with chloroform and methanol.

Notes
Moisture Sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383-63-1
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.077
MDL Number MFCD00000419
InChI Key STSCVKRWJPWALQ-UHFFFAOYSA-N
Synonym ethyl trifluoroacetate, trifluoroacetic acid ethyl ester, ethyltrifluoroacetate, acetic acid, trifluoro-, ethyl ester, trifluoroacetic acid, ethyl ester, ethyl trifluoroethanoate, unii-a6tzk6x11x, acetic acid, 2,2,2-trifluoro-, ethyl ester, a6tzk6x11x, ethyl ester of trifluoroacetic acid
PubChem CID 9794
IUPAC Name ethyl 2,2,2-trifluoroacetate
SMILES CCOC(=O)C(F)(F)F

Specifications

Melting Point <-78°C
Density 1.192
Boiling Point 60°C to 62°C
Flash Point −1°C (30°F)
Odor Sweet
Linear Formula CF3CO2CH2CH3
Refractive Index 1.307
Quantity 250 g
UN Number UN2924
Beilstein 1761411
Sensitivity Moisture sensitive
Solubility Information Slightly miscible with water. Miscible with chloroform and methanol.
Formula Weight 142.08
Percent Purity 99%
Chemical Name or Material Ethyl trifluoroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.