missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-3-(3-pyridyl)acrylate, 96%, Thermo Scientific™

Product Code. p-7032484
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456857 1 g 1g
15466857 5 g 5g
15476857 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15456857 Supplier Thermo Scientific Alfa Aesar Supplier No. H33964.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59607-99-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD02177443
InChI Key PIEQSBWGIODYPE-AATRIKPKSA-N
Synonym ethyl 3-3-pyridyl acrylate, e-ethyl 3-pyridin-3-yl acrylate, ethyl e-3-3-pyridinyl-2-propenoate, ethyl 2e-3-pyridin-3-yl prop-2-enoate, ethyl 3-pyridin-3-yl acrylate, ethyl 3-pyridineacrylate, 3-pyridineacrylic acid ethyl ester, ethyl 3-3-pyridyl propenoate, 2-propenoic acid, 3-3-pyridinyl-, ethyl ester, ethyl trans-3-3-pyridyl acrylate
PubChem CID 5371888
IUPAC Name ethyl (E)-3-pyridin-3-ylprop-2-enoate
SMILES CCOC(=O)C=CC1=CN=CC=C1

Specifications

Boiling Point 100°C to 102°C (0.075 mmHg)
Quantity 1 g
Formula Weight 177.2
Percent Purity 96%
Chemical Name or Material Ethyl trans-3-(3-pyridyl)acrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.