missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-2-(hydroxymethyl)cyclopropanecarboxylate, 97%, Thermo Scientific™

Product Code. 15478006
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478006 1 g 1g
15488006 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478006 Supplier Thermo Scientific Alfa Aesar Supplier No. H62920.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1026787-30-3
Molecular Formula C7H12O3
MDL Number MFCD18434533
Synonym trans-2-(Hydroxymethyl)cyclopropanecarboxylic acid ethyl ester

Specifications

Quantity 1 g
Formula Weight 144.16
Percent Purity 97%
Chemical Name or Material Ethyl trans-2-(hydroxymethyl)cyclopropanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.