missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10227120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10227120 1 g 1g
11955341 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10227120 Supplier Thermo Scientific Acros Supplier No. 265650010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28250-14-8
Molecular Formula C9H17NO2·HCl
Molecular Weight (g/mol) 207.7
InChI Key XMQSOBPCWYVZSW-SCLLHFNJSA-N
Synonym ethyl 1r,2r-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, trans-ethyl 2-aminocyclohexanecarboxylate hydrochloride, trans-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, 1r,2r-ethyl 2-aminocyclohexanecarboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride,
PubChem CID 2724642
IUPAC Name ethyl (1R,2R)-2-aminocyclohexane-1-carboxylate;hydrochloride
SMILES CCOC(=O)C1CCCCC1N.Cl

Specifications

Melting Point 152.0°C to 156.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water
Quantity 1 g
Formula Weight 207.7
Percent Purity 98%
Physical Form Granular Powder
Chemical Name or Material Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.