missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-4510360
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
28250-14-8
C9H17NO2·HCl
207.7
XMQSOBPCWYVZSW-SCLLHFNJSA-N
ethyl 1r,2r-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, trans-ethyl 2-aminocyclohexanecarboxylate hydrochloride, trans-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, 1r,2r-ethyl 2-aminocyclohexanecarboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride,
2724642
ethyl (1R,2R)-2-aminocyclohexane-1-carboxylate;hydrochloride
CCOC(=O)C1CCCCC1N.Cl
This item is not returnable. View return policy
28250-14-8
C9H17NO2·HCl
207.7
XMQSOBPCWYVZSW-SCLLHFNJSA-N
ethyl 1r,2r-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, trans-ethyl 2-aminocyclohexanecarboxylate hydrochloride, trans-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, 1r,2r-ethyl 2-aminocyclohexanecarboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride,
2724642
ethyl (1R,2R)-2-aminocyclohexane-1-carboxylate;hydrochloride
CCOC(=O)C1CCCCC1N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 152.0°C to 156.0°C
Color White
Assay Percent Range 97.5% min. (Argentometry)
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water
Formula Weight 207.7
Percent Purity 98%
Quantity 1 g
Physical Form Granular Powder
Chemical Name or Material Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'