missing translation for 'onlineSavingsMsg'
Learn More

Ethyl tiglate, 98%

Product Code. 11489892
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489892 25 g 25g
11499892 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489892 Supplier Thermo Scientific Alfa Aesar Supplier No. A12029.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5837-78-5 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 5837-78-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00015183
InChI Key OAPHLAAOJMTMLY-GQCTYLIASA-N
Synonym ethyl tiglate, tiglic acid, ethyl ester, ethyl 2-methylcrotonate, ethyl alpha-methylcrotonate, ethyl trans-2-methyl-2-butenoate, e-2-methyl-2-butenoic acid ethyl ester, ethyl e-2-methylcrotonate, ethyl trans-2-methylcrotonate, tiglic acid ethyl ester, ethyl 2-methylbut-2-enoate
PubChem CID 5281163
ChEBI CHEBI:4892
IUPAC Name ethyl (E)-2-methylbut-2-enoate
SMILES CCOC(=O)C(=CC)C

Spécification

Density 0.93
Boiling Point 156°C
Flash Point 46°C (114°F)
Linear Formula CH3CH=C(CH3)CO2CH2CH3
Refractive Index 1.434
Quantity 25 g
UN Number UN3272
Beilstein 1720895
Merck Index 14,9433
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Ethyl tiglate
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.